(R)-(+)-4-Chloro-3-hydroxybutyronitrile ≥96.0% (by GC)
Supplier: TCI America
C2763-1G
C2763-5G
TCC2763-1GEA
67.94
CAD
TCC2763-1G
TCC2763-5G
(R)-(+)-4-Chloro-3-hydroxybutyronitrile ≥96.0% (by GC)
(R)-(+)-4-Chloro-3-hydroxybutyronitrile
CAS Number: 84367-31-7
MDL Number: MFCD00273362
Molecular Formula: C4H6ClNO
Molecular Weight: 119.55
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 110
Flash Point (°C): 113
Specific Gravity (20/20): 1.25
Specific rotation [a]20/D: 13 deg (neat)
Formula:
C₄H₆ClNO Boiling Pt: 110 °C Density: 1.25 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00273362 CAS Number: 84367-31-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...