4-Chloro-3-(trifluoromethyl)benzaldehyde ≥97.0%

Supplier: TCI America

C2760-1G C2760-5G
TCC2760-1GEA 55.3 CAD
TCC2760-1G TCC2760-5G
4-Chloro-3-(trifluoromethyl)benzaldehyde ≥97.0%
4-Chloro-3-(trifluoromethyl)benzaldehyde

CAS Number: 34328-46-6
MDL Number: MFCD00040722
Molecular Formula: C8H4ClF3O
Molecular Weight: 208.56
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 54
Specific Gravity (20/20): 1.46

Formula: C₈H₄ClF₃O
Boiling Pt: 54 °C
Density: 1.46 (20 °C)
MDL Number: MFCD00040722
CAS Number: 34328-46-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca1eb847b57dbf1dac7df0057c255e18c3e6622c&vid=ca1eb847b57dbf1dac7df0057c255e18c3e6622c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR