N-Carbobenzoxy-6-aminohexanoic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
C2757-25G
C2757-5G
TCC2757-25GEA
391.84
CAD
TCC2757-25G
TCC2757-5G
N-Carbobenzoxy-6-aminohexanoic acid ≥98.0% (by HPLC, titration analysis)
N-Carbobenzoxy-6-aminohexanoic acid
CAS Number: 1947-00-8
MDL Number: MFCD00004423
Molecular Formula: C14H19NO4
Molecular Weight: 265.31
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 58
Flash Point (°C): 113
Storage Temperature: 0-10°C
Product Categories: Bioscience, Crosslinkers
Other Product Names / Synonyms: N-Cbz-6-aminohexanoic Acid, N-Carbobenzoxy-epsilon-aminocaproic Acid, N-Cbz-epsilon-aminocaproic Acid
Formula:
C₁₄H₁₉NO₄ Melting Pt: 58 °C Flash Pt: 113 °C |
MDL Number:
MFCD00004423 CAS Number: 1947-00-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...