(-)-Corey lactone-4-phenylbenzoate ≥98.0%
Supplier: TCI America
Synonyms:
(3aR,4S,5R,6aS)-4-(Hydroxymethyl)-2-oxohexahydro-2H-cyclopenta[b]furan-5-yl 4-Phenylbenzoate, (3aR,4S,5R,6aS)-Hexahydro-4-(hydroxymethyl)-5-(4-phenylbenzoyloxy)-2H-cyclopenta[b]furan-2-one, (3aR,4S,5R,6aS)-4-(Hydroxymethyl)-2-oxohexahydro-2H-cyclopenta[b]furan-5-yl Biphenyl-4-carboxylate
CAS Number: 31752-99-5
MDL Number: MFCD00078077
Molecular Formula: C21H20O5
Molecular Weight: 352.39
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 133
Specific rotation [a]20/D: -83 deg (C=1, CHCl3)
Storage Temperature: 0-10°C
Other Product Names / Synonyms: (3aR,4S,5R,6aS)-4-(Hydroxymethyl)-2-oxohexahydro-2H-cyclopenta[b]furan-5-yl 4-Phenylbenzoate, (3aR,4S,5R,6aS)-Hexahydro-4-(hydroxymethyl)-5-(4-phenylbenzoyloxy)-2H-cyclopenta[b]furan-2-one
Formula:
C₂₁H₂₀O₅ MW: 352.39 g/mol Melting Pt: 133 °C |
MDL Number:
MFCD00078077 CAS Number: 31752-99-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...