2-Chloro-5-methoxybenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

C2755-1G C2755-200MG
TCC2755-1GEA 319 CAD
TCC2755-1G TCC2755-200MG
2-Chloro-5-methoxybenzoic acid ≥98.0% (by GC, titration analysis)
2-Chloro-5-methoxybenzoic acid

CAS Number: 6280-89-3
MDL Number: MFCD00466261
Molecular Formula: C8H7ClO3
Molecular Weight: 186.59
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 174


Other Product Names / Synonyms: 6-Chloro-m-anisic Acid

Formula: C₈H₇ClO₃
Melting Pt: 174 °C
MDL Number: MFCD00466261
CAS Number: 6280-89-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ceffc2d1333d61a8aa8867ea57db1c091b2e01e&vid=2ceffc2d1333d61a8aa8867ea57db1c091b2e01e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR