(2-Chlorophenyl)acetylene 98,0

Supplier: TCI America

Synonyms: 1-Chloro-2-ethynylbenzene

C2750-1G C2750-5G
TCC2750-1GEA 52.14 CAD
TCC2750-1G TCC2750-5G
(2-Chlorophenyl)acetylene 98,0
(2-Chlorophenyl)acetylene

CAS Number: 873-31-4
MDL Number: MFCD00269951
Molecular Formula: C8H5Cl
Molecular Weight: 136.58
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 72
Flash Point (°C): 70
Specific Gravity (20/20): 1.13
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: 2-Chlorophenylacetylene

Formula: C₈H₅Cl
MW: 136.58
Boiling Pt: 72 °C
Density: 1.13 (20 °C)
Flash Pt: 70 °C
MDL Number: MFCD00269951
CAS Number: 873-31-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c014d1c5ac4e5518388d8bc4d3b6f92d02adbe5&vid=4db7d2c598d4e4f08ae3f3b85eb27fb5f4ab432c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR