4-Chloro-3-fluorophenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

C2749-1G C2749-5G
TCC2749-1GEA 36.34 CAD
TCC2749-1G TCC2749-5G
4-Chloro-3-fluorophenylboronic acid (contains varying amounts of Anhydride)
4-Chloro-3-fluorophenylboronic acid

CAS Number: 137504-86-0
MDL Number: MFCD01319010
Molecular Formula: C6H5BClFO2
Molecular Weight: 174.36
Form: Crystal
Color: White
Melting point (°C): 144


Other Product Names / Synonyms: 4-Chloro-3-fluorobenzeneboronic Acid

Formula: C₆H₅BClFO₂
Melting Pt: 144 °C
MDL Number: MFCD01319010
CAS Number: 137504-86-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5fba5ac2285f5954c59b517909e5dae17aaa35d&vid=c5fba5ac2285f5954c59b517909e5dae17aaa35d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR