5-Chloro-1,3-dimethyl-1H-pyrazole-4-carbaldehyde ≥98.0%

Supplier: TCI America

C2747-1G C2747-5G
TCC2747-1GEA 135.88 CAD
TCC2747-1G TCC2747-5G
5-Chloro-1,3-dimethyl-1H-pyrazole-4-carbaldehyde ≥98.0%
5-Chloro-1,3-dimethyl-1H-pyrazole-4-carbaldehyde

CAS Number: 27006-76-4
MDL Number: MFCD00052538
Molecular Formula: C6H7ClN2O
Molecular Weight: 158.59
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 79


Other Product Names / Synonyms: 5-Chloro-4-formyl-1,3-dimethylpyrazole

Formula: C₉H₁₁ClN₂O
MW: 158.59 g/mol
Melting Pt: 79 °C
MDL Number: MFCD00052538
CAS Number: 27006-76-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=135a83e16adb5cc6184f0633394154c96a6bdaed&vid=135a83e16adb5cc6184f0633394154c96a6bdaed&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR