Cetilistat ≥98.0% (by HPLC)

Supplier: TCI America

C2745-1G C2745-5G
TCC2745-1GEA 26.86 CAD
TCC2745-1G TCC2745-5G
Cetilistat ≥98.0% (by HPLC)
Cetilistat

CAS Number: 282526-98-1
MDL Number: MFCD09839697
Molecular Formula: C25H39NO3
Molecular Weight: 401.59
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 74


Product Categories: Bioscience, Obesity/Diabete Research Reagents, Inhibitors (for Pharmacological Research), Other Pharmaceutical Ingredients
Other Product Names / Synonyms: 2-Hexadecyloxy-6-methyl-4H-3,1-benzoxazin-4-one

Formula: C₂₅H₃₉NO₃
Melting Pt: 74 °C
MDL Number: MFCD09839697
CAS Number: 282526-98-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5719d1092a99fa3206336512eb440f0d066bc75e&vid=5719d1092a99fa3206336512eb440f0d066bc75e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR