5-Chloroisatoic anhydride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

C2743-25G C2743-5G
TCC2743-25GEA 488.22 CAD
TCC2743-25G TCC2743-5G
5-Chloroisatoic anhydride ≥98.0% (by HPLC, titration analysis)
5-Chloroisatoic anhydride

CAS Number: 4743-17-3
MDL Number: MFCD00006701
Molecular Formula: C8H4ClNO3
Molecular Weight: 197.57
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 272


Other Product Names / Synonyms: 6-Chloro-1,2-dihydro -4H-3,1-benzoxazine-2,4-dione

Formula: C₈H₄ClNO₃
Melting Pt: 272 °C
MDL Number: MFCD00006701
CAS Number: 4743-17-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98a2f834a927e61e42cae2ad73e5c2a28740c3a3&vid=98a2f834a927e61e42cae2ad73e5c2a28740c3a3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR