(S)-(+)-2-Chloromandelic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

C2738-25G C2738-5G
TCC2738-25GEA 1132.86 CAD
TCC2738-25G TCC2738-5G
(S)-(+)-2-Chloromandelic acid ≥98.0% (by GC, titration analysis)
(S)-(+)-2-Chloromandelic acid

CAS Number: 52950-19-3
MDL Number: MFCD00798437
Molecular Formula: C8H7ClO3
Molecular Weight: 186.59
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 121
Specific rotation [a]20/D: 125 deg (C=3, H2O)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-2'-Chloro-alpha-hydroxyphenylacetic Acid

Formula: C₈H₇ClO₃
Melting Pt: 121 °C
MDL Number: MFCD00798437
CAS Number: 52950-19-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff2954698a36ce82024126ccd6738178794a4fc2&vid=ff2954698a36ce82024126ccd6738178794a4fc2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR