3-(1-Cyanoethyl)benzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

C2730-25G C2730-5G
TCC2730-25GEA 102.7 CAD
TCC2730-25G TCC2730-5G
3-(1-Cyanoethyl)benzoic acid ≥98.0% (by GC, titration analysis)
3-(1-Cyanoethyl)benzoic acid

CAS Number: 5537-71-3
MDL Number: MFCD00002520
Molecular Formula: C10H9NO2
Molecular Weight: 175.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 148

Formula: C₁₀H₉NO₂
Melting Pt: 148 °C
MDL Number: MFCD00002520
CAS Number: 5537-71-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a02d7b802a34de2ee44b8e983774a9e3bd9482b7&vid=a02d7b802a34de2ee44b8e983774a9e3bd9482b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR