2-Chloro-4-(methylsulfonyl)benzoic acid ≥98.0%

Supplier: TCI America

Synonyms: 2-Chloro-4-methylsulfonylbenzoic acid, 2-Chloro-4-methylsulphonylbenzoic acid

C2726-25G C2726-5G
TCC2726-25GEA 123.24 CAD
TCC2726-25G TCC2726-5G
2-Chloro-4-(methylsulfonyl)benzoic acid ≥98.0%
2-Chloro-4-(methylsulphonyl)benzoic acid

CAS Number: 53250-83-2
MDL Number: MFCD00216496
Molecular Formula: C8H7ClO4S
Molecular Weight: 234.65
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 196

Formula: C₈H₇ClO₄S
Melting Pt: 196 °C
MDL Number: MFCD00216496
CAS Number: 53250-83-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5aa55802655571136899ceac9b230e899bba713e&vid=5aa55802655571136899ceac9b230e899bba713e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR