6-Chloropyrazine-2-carboxylic acid ≥96.0% (by HPLC, titration analysis)

Supplier: TCI America

C2725-1G C2725-5G
TCC2725-1GEA 158 CAD
TCC2725-1G TCC2725-5G
6-Chloropyrazine-2-carboxylic acid ≥96.0% (by HPLC, titration analysis)
6-Chloropyrazine-2-carboxylic acid

CAS Number: 23688-89-3
MDL Number: MFCD00837871
Molecular Formula: C5H3ClN2O2
Molecular Weight: 158.54
Purity/Analysis Method: >96.0% (HPLC,T)
Form: Crystal
Melting point (°C): 160

Formula: C₅H₃ClN₂O₂
MW: 158.54 g/mol
Melting Pt: 160 °C
MDL Number: MFCD00837871
CAS Number: 23688-89-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=678e034b4fe60db9665f6698f916c165689aa6f2&vid=678e034b4fe60db9665f6698f916c165689aa6f2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR