2-Chloro-4-formylpyridine ≥98.0%

Supplier: TCI America

Synonyms: 2-Chloropyridine-4-carboxaldehyde, 2-Chloroisonicotinaldehyde

C2723-1G C2723-5G
TCC2723-1GEA 126.4 CAD
TCC2723-1G TCC2723-5G
2-Chloro-4-formylpyridine ≥98.0%
2-Chloro-4-formylpyridine

CAS Number: 101066-61-9
MDL Number: MFCD06651557
Molecular Formula: C6H4ClNO
Molecular Weight: 141.55
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 49
Flash Point (°C): 110


Other Product Names / Synonyms: 2-Chloroisonicotinaldehyde, 2-Chloro-4-formylpyridine

Formula: C₆H₄ClNO
MW: 141.56
Melting Pt: 49 °C
Flash Pt: 110 °C
MDL Number: MFCD06651557
CAS Number: 101066-61-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91b720197b19699f8aa950c3ac8939bc8b5847e7&vid=91b720197b19699f8aa950c3ac8939bc8b5847e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR