1-Chloro-4-iodobutane ≥98.0% (by GC) stabilized

Supplier: TCI America

C2713-25G C2713-5G
TCC2713-25GEA 281.24 CAD
TCC2713-25G TCC2713-5G
1-Chloro-4-iodobutane ≥98.0% (by GC) stabilized
1-Chloro-4-iodobutane

(stabilized with Copper chip)
CAS Number: 10297-05-9
MDL Number: MFCD00039415
Molecular Formula: C4H8ClI
Molecular Weight: 218.46
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 67
Flash Point (°C): 93
Specific Gravity (20/20): 1.80


Other Product Names / Synonyms: 4-Chlorobutyl Iodide, 4-Iodobutyl Chloride

Formula: C₄H₈ClI
Boiling Pt: 67 °C
Density: 1.80 (20 °C)
Flash Pt: 93 °C
MDL Number: MFCD00039415
CAS Number: 10297-05-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9241f05216df2d54dd7c64aa5d18a07ed5fd4553&vid=9241f05216df2d54dd7c64aa5d18a07ed5fd4553&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR