1,4-Dibenzyl-L-aspartate-p-toluenesulfonate ≥97.0%

Supplier: TCI America

Synonyms: H-Asp(OBzl)-Obzl.TosOH, L-Aspartic Acid Dibenzyl Ester p-Toluenesulphonate Salt, L-Aspartic acid dibenzyl ester p-toluenesulfonate, L-Aspartic acid dibenzyl ester p-toluenesulphonate salt Dibenzyl (2S)-2-aminobutanedioate 4-toluenesulphonate, H-Asp(OBzl)-OBzl.Tos-OH, L-Aspartic Acid 1,4-Dibenzyl Ester p-Toluenesulphonate, H-Asp(OBzl)-OBzl·TosOH, L-Aspartic acid dibenzyl ester p-toluenesulphonate, L-Aspartic Acid 1,4-Dibenzyl Ester p-Toluenesulfonate, L-Aspartic Acid Dibenzyl Ester p-Toluenesulfonate Salt, L-Aspartic acid dibenzyl ester p-toluenesulfonate salt Dibenzyl (2S)-2-aminobutanedioate 4-toluenesulfonate

C2711-25G C2711-5G
TCC2711-25GEA 423.44 CAD
TCC2711-25G TCC2711-5G
1,4-Dibenzyl-L-aspartate-p-toluenesulfonate ≥97.0%
1,4-Dibenzyl-L-aspartate-p-toluenesulfonate

CAS Number: 2886-33-1
MDL Number: MFCD00065188
Molecular Formula: C18H19NO4
Molecular Weight: 485.55
Purity/Analysis Method: >97.0% (HPLC,N,T)
Form: Crystal
Melting point (°C): 159
Specific rotation [a]20/D: 9 deg (C=1, CHCl3)


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: L-Aspartic Acid 1,4-Dibenzyl Ester p-Toluenesulfonate, H-Asp(OBzl)-OBzl.Tos-OH

Formula: C₁₈H₁₉NO₄
MW: 485.55 g/mol
Melting Pt: 159 °C
MDL Number: MFCD00065188
CAS Number: 2886-33-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=240a9bc735655f3b26694671b28920e0fe8ccf19&vid=240a9bc735655f3b26694671b28920e0fe8ccf19&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,N,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR