1-Cyclopropyl-2-(2-fluorophenyl)ethanone ≥98.0% (by GC)

Supplier: TCI America

C2710-25G C2710-5G
TCC2710-25GEA 350.76 CAD
TCC2710-25G TCC2710-5G
1-Cyclopropyl-2-(2-fluorophenyl)ethanone ≥98.0% (by GC)
1-Cyclopropyl-2-(2-fluorophenyl)ethanone

CAS Number: 150322-73-9
MDL Number: MFCD10687164
Molecular Formula: C11H11FO
Molecular Weight: 178.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 61
Specific Gravity (20/20): 1.12

Formula: C₁₁H₁₁FO
Boiling Pt: 61 °C
Density: 1.12 (20 °C)
MDL Number: MFCD10687164
CAS Number: 150322-73-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f5c4ddba6f4d03c6d40499f19f0e21e41653951&vid=0f5c4ddba6f4d03c6d40499f19f0e21e41653951&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR