N-(Carbobenzoxy)-1H-pyrazole-1-carboxamidine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

C2709-1G C2709-5G
TCC2709-1GEA 48.98 CAD
TCC2709-1G TCC2709-5G
N-(Carbobenzoxy)-1H-pyrazole-1-carboxamidine ≥98.0% (by HPLC, titration analysis)
N-(Carbobenzoxy)-1H-pyrazole-1-carboxamidine

CAS Number: 152120-62-2
MDL Number: MFCD00274657
Molecular Formula: C12H12N4O2
Molecular Weight: 244.25
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 111


Product Categories: Synthetic Organic Chemistry, Guanidinylation
Other Product Names / Synonyms: N-(Benzyloxycarbonyl)-1H-pyrazole-1-carboxamidine, N-Cbz-1H-pyrazole-1-carboxamidine, 1-[N-(Carbobenzoxy)amidino]pyrazole, 1-(N-Cbz-amidino)pyrazole

Formula: C₁₂H₁₂N₄O₂
Melting Pt: 111 °C
MDL Number: MFCD00274657
CAS Number: 152120-62-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0603abeae56cf57faddbc54401d807a428c0805&vid=a0603abeae56cf57faddbc54401d807a428c0805&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR