3-Chloro-5-hydroxybenzonitrile ≥95.0% (by GC)
Supplier: TCI America
C2708-1G
C2708-5G
TCC2708-1GEA
133.83
CAD
TCC2708-1G
TCC2708-5G
3-Chloro-5-hydroxybenzonitrile ≥95.0% (by GC)
3-Chloro-5-hydroxybenzonitrile
CAS Number: 473923-97-6
MDL Number: MFCD11226540
Molecular Formula: C7H4ClNO
Molecular Weight: 153.57
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 170
Other Product Names / Synonyms: 3-Chloro-5-cyanophenol
Formula:
C₇H₄ClNO Melting Pt: 170 °C |
MDL Number:
MFCD11226540 CAS Number: 473923-97-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...