4-Chloro-1-fluoro-2-nitrobenzene ≥98.0% (by GC)

Supplier: TCI America

C2704-25G C2704-5G
TCC2704-25GEA 271.76 CAD
TCC2704-25G TCC2704-5G
4-Chloro-1-fluoro-2-nitrobenzene ≥98.0% (by GC)
4-Chloro-1-fluoro-2-nitrobenzene

CAS Number: 345-18-6
MDL Number: MFCD00069418
Molecular Formula: C6H3ClFNO2
Molecular Weight: 175.54
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 237
Melting point (°C): 10
Specific Gravity (20/20): 1.48


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Chloro-1-fluoro-2-nitrobenzene

Formula: C₆H₃ClFNO₂
Boiling Pt: 237 °C
Melting Pt: 10 °C
Density: 1.48 (20 °C)
MDL Number: MFCD00069418
CAS Number: 345-18-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca2cbce28fc38b92a6d445e1a502d2634d33ca42&vid=ca2cbce28fc38b92a6d445e1a502d2634d33ca42&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR