2-Chloro-4,6-diphenyl-1,3,5-triazine ≥98.0% (by GC)

Supplier: TCI America

C2703-25G C2703-5G
TCC2703-25GEA 319.16 CAD
TCC2703-25G TCC2703-5G
2-Chloro-4,6-diphenyl-1,3,5-triazine ≥98.0% (by GC)
2-Chloro-4,6-diphenyl-1,3,5-triazine

CAS Number: 3842-55-5
MDL Number: MFCD05738885
Molecular Formula: C15H10ClN3
Molecular Weight: 267.72
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 140
Specific Gravity (20/20): 1.06

Formula: C15H10ClN3
MW: 267.72 g/mol
Melting Pt: 140 °C
Density: 1.06 (20 °C)
MDL Number: MFCD05738885
CAS Number: 3842-55-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1f6c1a4cd03971c49fb6f71dd06e8c35c9d2c1a&vid=b1f6c1a4cd03971c49fb6f71dd06e8c35c9d2c1a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR