(S)-Benzyl(2-oxotetrahydrofuran-3-yl)carbamate 98,0
Supplier: TCI America
Synonyms:
N-Cbz-L-homoserine Lactone, (S)-alpha-(Carbobenzoxyamino)-gamma-butyrolactone, Cbz-Hse-Lactone, (S)-alpha-(Benzyloxycarbonylamino)-gamma-butyrolactone, N-Benzyloxycarbonyl-L-homoserine Lactone, (S)-Benzyl (2-oxotetrahydrofuran-3-yl)carbamate, N-Carbobenzoxy-L-homoserine Lactone, (S)-alpha-(Cbz-amino)-gamma-butyrolactone
CAS Number: 35677-89-5
MDL Number: MFCD00674555
Molecular Formula: C12H13NO4
Molecular Weight: 235.24
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 129
Specific rotation [a]20/D: -32 deg (C=1, MeOH)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Chiral Building Blocks
Other Product Names / Synonyms: N-Cbz-L-homoserine Lactone, (S)-alpha-(Carbobenzoxyamino)-gamma-butyrolactone, (S)-alpha-(Cbz-amino)-gamma-butyrolactone
Formula:
C₁₂H₁₃NO₄ MW: 235.2400 g/mol Melting Pt: 129 °C |
MDL Number:
MFCD00674555 CAS Number: 35677-89-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...