2-Chloro-5-methoxypyrimidine ≥98.0%

Supplier: TCI America

C2693-1G C2693-5G
TCC2693-1GEA 61.62 CAD
TCC2693-1G TCC2693-5G
2-Chloro-5-methoxypyrimidine ≥98.0%
2-Chloro-5-methoxypyrimidine

CAS Number: 22536-65-8
MDL Number: MFCD08702770
Molecular Formula: C5H5ClN2O
Molecular Weight: 144.56
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 77

Formula: C₅H₅ClN₂O
MW: 144.56 g/mol
Melting Pt: 77 °C
MDL Number: MFCD08702770
CAS Number: 22536-65-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ccfb112917f9741b3aada59c87209b7c56bb710&vid=1ccfb112917f9741b3aada59c87209b7c56bb710&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR