3-Cyano-4-fluorobenzaldehyde ≥98.0% (by GC)

Supplier: TCI America

C2692-1G C2692-5G
TCC2692-1GEA 52.93 CAD
TCC2692-1G TCC2692-5G
3-Cyano-4-fluorobenzaldehyde ≥98.0% (by GC)
3-Cyano-4-fluorobenzaldehyde

CAS Number: 218301-22-5
MDL Number: MFCD01863558
Molecular Formula: C8H4FNO
Molecular Weight: 149.12
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 85


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Fluoro-5-formylbenzonitrile

Formula: C₈H₄FNO
Melting Pt: 85 °C
MDL Number: MFCD01863558
CAS Number: 218301-22-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d93270650e0191a38260ae53591e007da0268dd&vid=4d93270650e0191a38260ae53591e007da0268dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR