2-(((Benzyloxy)carbonyl)amino)-2-hydroxyacetic acid ≥98.0%

Supplier: TCI America

Synonyms: Z-2-hydroxyglycine, 2-(Benzyloxycarbonylamino)-2-hydroxyacetic acid, 2-(Carbobenzoxyamino)-2-hydroxyacetic acid, N-Benzyloxycarbonyl-2-hydroxyglycine, N-Carbobenzoxy-2-hydroxyglycine, 2-(Cbz-amino)-2-hydroxyacetic acid

C2681-1G C2681-5G
TCC2681-1GEA 45.82 CAD
TCC2681-1G TCC2681-5G
2-(((Benzyloxy)carbonyl)amino)-2-hydroxyacetic acid ≥98.0%
2-(((Benzyloxy)carbonyl)amino)-2-hydroxyacetic acid

CAS Number: 56538-57-9
MDL Number: MFCD02179279
Molecular Formula: C10H11NO5
Molecular Weight: 225.20
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 189


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: 2-(Cbz-amino)-2-hydroxyacetic Acid, N-Carbobenzoxy-2-hydroxyglycine, Z-2-hydroxyglycine

Formula: C10H11NO5
MW: 225.2 g/mol
Melting Pt: 189 °C
MDL Number: MFCD02179279
CAS Number: 56538-57-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4060e8b3f14809040120c13a6bafc2f6b67af482&vid=4060e8b3f14809040120c13a6bafc2f6b67af482&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR