1-(Benzyloxycarbonylamino)cyclopropyl-1-carboxylic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
C2680-1G
C2680-5G
TCC2680-1GEA
52.14
CAD
TCC2680-1G
TCC2680-5G
1-(Benzyloxycarbonylamino)cyclopropyl-1-carboxylic acid ≥98.0% (by HPLC, titration analysis)
1-(Benzyloxycarbonylamino)cyclopropyl-1-carboxylic acid
CAS Number: 84677-06-5
MDL Number: MFCD00083285
Molecular Formula: C12H13NO4
Molecular Weight: 235.24
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 159
Storage Temperature: 0-10°C
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: 1-(Cbz-amino)cyclopropanecarboxylic Acid
Formula:
C₁₂H₁₃NO₄ Melting Pt: 159 °C |
MDL Number:
MFCD00083285 CAS Number: 84677-06-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...