1,2,3,4-Cyclobutanetetracarboxdiimide ≥98.0% (by total nitrogen basis)
Supplier: TCI America
C2679-1G
C2679-200MG
TCC2679-1GEA
1489.31
CAD
TCC2679-1G
TCC2679-200MG
1,2,3,4-Cyclobutanetetracarboxdiimide ≥98.0% (by total nitrogen basis)
1,2,3,4-Cyclobutanetetracarboxdiimide
CAS Number: 4415-88-7
Molecular Formula: C8H6N2O4
Molecular Weight: 194.15
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 350
Formula:
C₈H₆N₂O₄ Melting Pt: 350 °C |
CAS Number:
4415-88-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...