4,6-Dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile ≥98.0%

Supplier: TCI America

C2672-25G C2672-5G
TCC2672-25GEA 145.36 CAD
TCC2672-25G TCC2672-5G
4,6-Dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile ≥98.0%
4,6-Dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile

CAS Number: 769-28-8
MDL Number: MFCD00006269
Molecular Formula: C8H8N2O
Molecular Weight: 148.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 290


Other Product Names / Synonyms: 2-Hydroxy-4,6-dimethylnicotinonitrile, 2-Hydroxy-4,6-dimethylpyridine-3-carbonitrile

Formula: C₈H₈N₂O
MW: 148.17 g/mol
Melting Pt: 290 °C
MDL Number: MFCD00006269
CAS Number: 769-28-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=20a744ff5879424e0299116dfa4ce69d2c43fd5c&vid=20a744ff5879424e0299116dfa4ce69d2c43fd5c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR