Cyclopenta[fg]tetracene-1,2-dione

Supplier: TCI America

C2669-1G C2669-200MG
TCC2669-1GEA 968.54 CAD
TCC2669-1G TCC2669-200MG
Cyclopenta[fg]tetracene-1,2-dione
Cyclopenta[fg]tetracene-1,2-dione

CAS Number: 1334510-66-5
Molecular Formula: C20H10O2
Molecular Weight: 282.30
Form: Crystal
Color: Deep Blue


Other Product Names / Synonyms: Cyclopenta[fg]naphthacene-1,2-dione

Formula: C₂₀H₁₀O₂
CAS Number: 1334510-66-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=798637053e2d227d0f6526fd1d7b3462fafb48bd&vid=798637053e2d227d0f6526fd1d7b3462fafb48bd&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR