Ciprofibrate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

C2667-1G C2667-5G
TCC2667-1GEA 268.6 CAD
TCC2667-1G TCC2667-5G
Ciprofibrate ≥98.0% (by GC, titration analysis)
Ciprofibrate

CAS Number: 52214-84-3
MDL Number: MFCD00467135
Molecular Formula: C13H14Cl2O3
Molecular Weight: 289.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 118
Storage Temperature: 0-10°C


Product Categories: Bioscience, Antilipemic Ingredients
Other Product Names / Synonyms: 2-[4-(2,2-Dichlorocyclopropyl)phenoxy]-2-methylpropionic Acid

Formula: C₁₃H₁₄Cl₂O₃
Melting Pt: 118 °C
MDL Number: MFCD00467135
CAS Number: 52214-84-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b347cab1f14795ad36f94e2286642b133b362758&vid=b347cab1f14795ad36f94e2286642b133b362758&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR