5-Chloro-2-methylphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

C2665-1G C2665-5G
TCC2665-1GEA 71.1 CAD
TCC2665-1G TCC2665-5G
5-Chloro-2-methylphenylboronic acid (contains varying amounts of Anhydride)
5-Chloro-2-methylphenylboronic acid

CAS Number: 148839-33-2
MDL Number: MFCD03411939
Molecular Formula: C7H8BClO2
Molecular Weight: 170.40
Form: Crystal
Color: White
Melting point (°C): 186


Other Product Names / Synonyms: 5-Chloro-2-methylbenzeneboronic Acid, 5-Chloro-o-tolylboronic Acid

Formula: C₇H₈BClO₂
Melting Pt: 186 °C
MDL Number: MFCD03411939
CAS Number: 148839-33-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7dee74150774fb8bc7314fc2ba1cf53128c17dab&vid=7dee74150774fb8bc7314fc2ba1cf53128c17dab&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR