3-Chloro-4-ethoxyphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: (3-Chloro-4-ethoxyphenyl)boronic acid

C2664-1G C2664-5G
TCC2664-1GEA 54.51 CAD
TCC2664-1G TCC2664-5G
3-Chloro-4-ethoxyphenylboronic acid (contains varying amounts of Anhydride)
3-Chloro-4-ethoxyphenylboronic acid

CAS Number: 279261-81-3
MDL Number: MFCD03701556
Molecular Formula: C8H10BClO3
Molecular Weight: 200.43
Form: Crystal
Color: White
Melting point (°C): 239


Other Product Names / Synonyms: 3-Chloro-4-ethoxybenzeneboronic Acid

Formula: C₈H₁₀BClO₃
MW: 200.4300 g/mol
Melting Pt: 239 °C
MDL Number: MFCD03701556
CAS Number: 279261-81-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7f58eca2a4cc9ec95d90443a5a1bfbd30a3a8e18&vid=7f58eca2a4cc9ec95d90443a5a1bfbd30a3a8e18&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR