Methoxy(cyclooctadiene)iridium(I) dimer

Supplier: TCI America

Synonyms: Di-mu-methoxobis(1,5-cyclooctadiene)diiridium(I), Bis(1,5-cyclooctadiene)di-mu-methoxydiiridium(I), (1,5-Cyclooctadiene)(methoxy)iridium(I) dimer, Bis(1,5-cyclooctadiene)dimethoxydiiridium

C2662-1G C2662-200MG
TCC2662-1GEA 1186.58 CAD
TCC2662-1G TCC2662-200MG
Methoxy(cyclooctadiene)iridium(I) dimer
Methoxy(cyclooctadiene)iridium(I) dimer

CAS Number: 12148-71-9
MDL Number: MFCD08459360
Molecular Formula: C18H30Ir2O2
Molecular Weight: 662.87
Form: Crystal
Color: Pale Yellow
Melting point (°C): 179
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Bis(1,5-cyclooctadiene)di-mu-methoxydiiridium(I)

Formula: C₁₈H₃₀Ir₂O₂
MW: 662.87 g/mol
Melting Pt: 179 °C
MDL Number: MFCD08459360
CAS Number: 12148-71-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=015650b0f902db1fe11a60d52d0fd82bab3e4e2c&vid=015650b0f902db1fe11a60d52d0fd82bab3e4e2c&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR