3-Cyano-6-isopropylchromone ≥98.0% (by GC)

Supplier: TCI America

C2661-1G C2661-5G
TCC2661-1GEA 178.54 CAD
TCC2661-1G TCC2661-5G
3-Cyano-6-isopropylchromone ≥98.0% (by GC)
3-Cyano-6-isopropylchromone

CAS Number: 50743-32-3
MDL Number: MFCD00191961
Molecular Formula: C13H11NO2
Molecular Weight: 213.24
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 119


Other Product Names / Synonyms: 6-Isopropyl-4-oxo-4H-1-benzopyran-3-carbonitrile, 3-Cyano-6-isopropylchromone

Formula: C₁₃H₁₁NO₂
Melting Pt: 119 °C
MDL Number: MFCD00191961
CAS Number: 50743-32-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=510586669b425d7577c6d1b284470b9950e6ec5d&vid=510586669b425d7577c6d1b284470b9950e6ec5d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR