3-Cyclohexyl-D-alanine hydrate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: D-CHA-OH Hydrate, (R)-2-Amino-3-cyclohexylpropanoic acid hydrate(1:x)

C2659-1G C2659-5G
TCC2659-1GEA 183.44 CAD
TCC2659-1G TCC2659-5G
3-Cyclohexyl-D-alanine hydrate ≥98.0% (by HPLC, titration analysis)
3-Cyclohexyl-D-alanine hydrate

CAS Number: 213178-94-0
MDL Number: MFCD00217056
Molecular Formula: C9H17NO2
Molecular Weight: 171.24
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: -12 deg (C=1, 1mol/L HCl)

Formula: C₉H₁₇NO₂
MW: 189.25 g/mol
MDL Number: MFCD00217056
CAS Number: 213178-94-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=09259fc436f476ac3d4ce1c0d83612fe6da4183c&vid=09259fc436f476ac3d4ce1c0d83612fe6da4183c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR