Cyclopentylmethanol ≥98.0% (by GC)

Supplier: TCI America

C2658-25G C2658-5G
TCC2658-25GEA 423.44 CAD
TCC2658-25G TCC2658-5G
Cyclopentylmethanol ≥98.0% (by GC)
Cyclopentylmethanol

CAS Number: 3637-61-4
MDL Number: MFCD00001384
Molecular Formula: C6H12O
Molecular Weight: 100.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 163
Flash Point (°C): 62
Specific Gravity (20/20): 0.94


Other Product Names / Synonyms: Cyclopentylmethanol

Formula: C₆H₁₂O
Boiling Pt: 163 °C
Density: 0.94 (20 °C)
Flash Pt: 62 °C
MDL Number: MFCD00001384
CAS Number: 3637-61-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=10b7c8ef6f45d0de22cab1c32110d7197a69392c&vid=10b7c8ef6f45d0de22cab1c32110d7197a69392c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR