4-Chlorosalicylaldehyde ≥95.0% (by GC, titration analysis)

Supplier: TCI America

C2653-1G C2653-5G
TCC2653-1GEA 124.82 CAD
TCC2653-1G TCC2653-5G
4-Chlorosalicylaldehyde ≥95.0% (by GC, titration analysis)
4-Chlorosalicylaldehyde

CAS Number: 2420-26-0
MDL Number: MFCD06252499
Molecular Formula: C7H5ClO2
Molecular Weight: 156.57
Purity/Analysis Method: >95.0% (GC,T)
Form: Crystal
Melting point (°C): 47


Other Product Names / Synonyms: 4-Chloro-2-hydroxybenzaldehyde

Formula: C₇H₅ClO₂
Melting Pt: 47 °C
MDL Number: MFCD06252499
CAS Number: 2420-26-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c59df6516824e5cc37595c743d10ff4dfdd6721d&vid=c59df6516824e5cc37595c743d10ff4dfdd6721d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR