ɑ-(4-Chlorophenyl)-2-pyridinemethanol 98,0

Supplier: TCI America

Synonyms: (4-Chlorophenyl)(pyridin-2-yl)methanol, 2-[(4-Chlorophenyl)(hydroxy)methyl]pyridine, (4-chlorophenyl)(pyridin-2-yl)methanol, α-(4-Chlorophenyl)-2-pyridinemethanol

C2646-1G C2646-5G
TCC2646-1GEA 319 CAD
TCC2646-1G TCC2646-5G
ɑ-(4-Chlorophenyl)-2-pyridinemethanol 98,0
alpha-(4-Chlorophenyl)-2-pyridinemethanol

CAS Number: 27652-89-7
MDL Number: MFCD00023493
Molecular Formula: C12H10ClNO
Molecular Weight: 219.67
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 84

Formula: C₁₂H₁₀ClNO
MW: 219.67 g/mol
Melting Pt: 84 °C
MDL Number: MFCD00023493
CAS Number: 27652-89-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=817935057cf71563bd7dd3769b89457ddbc14ea4&vid=817935057cf71563bd7dd3769b89457ddbc14ea4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR