2'-Chloro-4'-fluoroacetophenone ≥97.0%

Supplier: TCI America

Synonyms: 1-(2-Chloro-4-fluorophenyl)ethanone

C2643-1G C2643-5G
TCC2643-1GEA 58.46 CAD
TCC2643-1G TCC2643-5G
2'-Chloro-4'-fluoroacetophenone ≥97.0%
2'-Chloro-4'-fluoroacetophenone

CAS Number: 700-35-6
MDL Number: MFCD00042202
Molecular Formula: C8H6ClFO
Molecular Weight: 172.58
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 110
Specific Gravity (20/20): 1.30

Formula: C₈H₆ClFO
MW: 172.59 g/mol
Boiling Pt: 110 °C
Density: 1.30 (20 °C)
MDL Number: MFCD00042202
CAS Number: 700-35-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e9b853ea01df1c40687fbd4731bb24cfbc9e915&vid=7e9b853ea01df1c40687fbd4731bb24cfbc9e915&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR