2-Chloro-4-methylpyrimidine ≥98.0% (by GC)
Supplier: TCI America
C2631-25G
C2631-5G
TCC2631-25GEA
262.28
CAD
TCC2631-25G
TCC2631-5G
2-Chloro-4-methylpyrimidine ≥98.0% (by GC)
2-Chloro-4-methylpyrimidine
CAS Number: 13036-57-2
MDL Number: MFCD00054434
Molecular Formula: C5H5ClN2
Molecular Weight: 128.56
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 94
Melting point (°C): 49
Formula:
C₅H₅ClN₂ Boiling Pt: 94 °C Melting Pt: 49 °C |
MDL Number:
MFCD00054434 CAS Number: 13036-57-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...