Cyclopropanesulfonamide ≥98.0% (by GC)

Supplier: TCI America

C2628-1G C2628-5G
TCC2628-1GEA 33.18 CAD
TCC2628-1G TCC2628-5G
Cyclopropanesulfonamide ≥98.0% (by GC)
Cyclopropanesulphonamide

CAS Number: 154350-29-5
MDL Number: MFCD08705286
Molecular Formula: C3H7NO2S
Molecular Weight: 121.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 108

Formula: C₃H₇NO₂S
Melting Pt: 108 °C
MDL Number: MFCD08705286
CAS Number: 154350-29-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83a01817248b464f50a3353237908ae52c27163c&vid=83a01817248b464f50a3353237908ae52c27163c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR