4-Cyano-4'-octyloxybiphenyl ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 4'-Octyloxy-[1,1'-biphenyl]-4-carbonitrile

C2618-25G C2618-5G
TCC2618-25GEA 900.92 CAD
TCC2618-25G TCC2618-5G
4-Cyano-4'-octyloxybiphenyl ≥98.0% (by GC)
4-Cyano-4'-octyloxybiphenyl

CAS Number: 52364-73-5
MDL Number: MFCD00075145
Molecular Formula: C21H25NO
Molecular Weight: 307.44
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 54
Flash Point (°C): 113


Product Categories: Materials Chemistry, Liquid Crystal Materials (Optically Functional Materials)
Other Product Names / Synonyms: 4'-n-Octyloxy-4-biphenylcarbonitrile, 8OCB

Formula: C₂₁H₂₅NO
MW: 307.44 g/mol
Melting Pt: 54 °C
Flash Pt: 113 °C
MDL Number: MFCD00075145
CAS Number: 52364-73-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dffd2d29676e1dc7456d27bd3a29a0321d8252a2&vid=dffd2d29676e1dc7456d27bd3a29a0321d8252a2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR