3-Chloro-1-fluoro-4-nitrobenzene ≥98.0% (by GC)

Supplier: TCI America

C2615-25G C2615-5G
TCC2615-25GEA 440.82 CAD
TCC2615-25G TCC2615-5G
3-Chloro-1-fluoro-4-nitrobenzene ≥98.0% (by GC)
3-Chloro-1-fluoro-4-nitrobenzene

CAS Number: 2106-50-5
MDL Number: MFCD03412200
Molecular Formula: C6H3ClFNO2
Molecular Weight: 175.54
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 71
Melting point (°C): 37


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Chloro-4-fluoro-1-nitrobenzene

Formula: C₆H₃ClFNO₂
Boiling Pt: 71 °C
Melting Pt: 37 °C
MDL Number: MFCD03412200
CAS Number: 2106-50-5
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9675336075703fec99e9d563128400fe0aefc8ca&vid=b8783674d8228451c39218751e38e6b3a98bbad1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR