N-Carbobenzoxy-S-benzyl-L-cysteine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

C2614-25G C2614-5G
TCC2614-25GEA 167.48 CAD
TCC2614-25G TCC2614-5G
N-Carbobenzoxy-S-benzyl-L-cysteine ≥98.0% (by HPLC, titration analysis)
N-Carbobenzoxy-S-benzyl-L-cysteine

CAS Number: 3257-18-9
MDL Number: MFCD00004785
Molecular Formula: C18H19NO4S
Molecular Weight: 345.41
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 98
Specific rotation [a]20/D: -46 deg (C=4, EtOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Cbz-S-benzyl-L-cysteine, Z-Cys(Bzl)-OH

Formula: C₁₈H₁₉NO₄S
Melting Pt: 98 °C
MDL Number: MFCD00004785
CAS Number: 3257-18-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9835821c0bd80f1e56af5624cdbf951b2c2d0a0f&vid=9835821c0bd80f1e56af5624cdbf951b2c2d0a0f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR