Copper(I) hexafluoro-2,4-pentanedionate-1,5-cyclooctadiene complex ≥95.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 1,5-Cyclooctadiene(hexafluoro-2,4-pentanedionato)copper(I), Copper(I) Hexafluoro-2,4-pentanedionate 1,5-Cyclooctadiene Complex

C2610-1G C2610-5G
TCC2610-1GEA 206.98 CAD
TCC2610-1G TCC2610-5G
Copper(I) hexafluoro-2,4-pentanedionate-1,5-cyclooctadiene complex ≥95.0% (by titrimetric analysis)
Copper(I) hexafluoro-2,4-pentanedionate-1,5-cyclooctadiene complex

CAS Number: 86233-74-1
MDL Number: MFCD00156517
Molecular Formula: C13H13CuF6O2
Molecular Weight: 378.78
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 100
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts
Other Product Names / Synonyms: 1,5-Cyclooctadiene(hexafluoro-2,4-pentanedionato)copper(I)

Formula: C₁₃H₁₃CuF₆O₂
MW: 378.78 g/mol
Melting Pt: 100 °C
MDL Number: MFCD00156517
CAS Number: 86233-74-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b793f8c897d01f051fdd878e7e163961f85b1d92&vid=b793f8c897d01f051fdd878e7e163961f85b1d92&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR