3-Cyano-4-methylpyridine ≥98.0%

Supplier: TCI America

Synonyms: 4-Methylpyridine-3-carbonitrile, 3-Cyano-4-picoline, 4-Methylnicotinonitrile

C2608-1G C2608-5G
TCC2608-1GEA 86.9 CAD
TCC2608-1G TCC2608-5G
3-Cyano-4-methylpyridine ≥98.0%
3-Cyano-4-methylpyridine

CAS Number: 5444-01-9
MDL Number: MFCD00234272
Molecular Formula: C7H6N2
Molecular Weight: 118.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 111
Melting point (°C): 47


Other Product Names / Synonyms: 4-Methyl-3-pyridinecarbonitrile, 4-Methylnicotinonitrile

Formula: C₇H₆N₂
MW: 118.14 g/mol
Boiling Pt: 111 °C
Melting Pt: 47 °C
MDL Number: MFCD00234272
CAS Number: 5444-01-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f5b6bbedf3df51d2080e7511957ef69e6345688a&vid=f5b6bbedf3df51d2080e7511957ef69e6345688a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR