2-Chloro-4-(dimethylamino)benzaldehyde ≥98.0% (by GC, total nitrogen)

Supplier: TCI America

C2606-1G C2606-5G
TCC2606-1GEA 91.64 CAD
TCC2606-1G TCC2606-5G
2-Chloro-4-(dimethylamino)benzaldehyde ≥98.0% (by GC, total nitrogen)
2-Chloro-4-(dimethylamino)benzaldehyde

CAS Number: 1424-66-4
MDL Number: MFCD00021051
Molecular Formula: C9H10ClNO
Molecular Weight: 183.64
Purity/Analysis Method: >98.0% (GC,N)
Form: Crystal
Melting point (°C): 82

Formula: C₉H₁₀ClNO
Melting Pt: 82 °C
MDL Number: MFCD00021051
CAS Number: 1424-66-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d9bf9bf953a9f5b217c4814cc2891bcad5a6f36&vid=0d9bf9bf953a9f5b217c4814cc2891bcad5a6f36&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR