(-)-Corey lactone benzoate ≥98.0% (by HPLC)

Supplier: TCI America

C2605-1G C2605-200MG
TCC2605-1GEA 480.95 CAD
TCC2605-1G TCC2605-200MG
(-)-Corey lactone benzoate ≥98.0% (by HPLC)
(-)-Corey lactone benzoate

CAS Number: 39746-00-4
MDL Number: MFCD00674102
Molecular Formula: C15H16O5
Molecular Weight: 276.29
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 120
Specific rotation [a]20/D: -75 deg (C=1, CHCl3)


Other Product Names / Synonyms: (3aR,4S,5R,6aS)-4-(Hydroxymethyl)-2-oxohexahydro-2H-cyclopenta[b]furan-5-yl Benzoate, (3aR,4S,5R,6aS)-5-(Benzoyloxy)hexahydro-4-(hydroxymethyl)-2H-cyclopenta[b]furan-2-one

Formula: C₁₅H₁₆O₅
Melting Pt: 120 °C
MDL Number: MFCD00674102
CAS Number: 39746-00-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=555f75a9c0f2343ace01e051c7b77ccbf31af1e7&vid=555f75a9c0f2343ace01e051c7b77ccbf31af1e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR