4-Chloro-2-iodobenzoic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

C2604-1G C2604-5G
TCC2604-1GEA 66.36 CAD
TCC2604-1G TCC2604-5G
4-Chloro-2-iodobenzoic acid ≥98.0% (by titrimetric analysis)
4-Chloro-2-iodobenzoic acid

CAS Number: 13421-13-1
MDL Number: MFCD00060676
Molecular Formula: C7H4ClIO2
Molecular Weight: 282.46
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 164

Formula: C₇H₄ClIO₂
Melting Pt: 164 °C
MDL Number: MFCD00060676
CAS Number: 13421-13-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b22a93c2c2cfaa95f8670582722607512f1c351b&vid=b22a93c2c2cfaa95f8670582722607512f1c351b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR